
Isonicotinamide ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1453-82-3
:
Formula:C₆H₆N₂O
MDL Number:MFCD00006432
:122.13 g/mol
Boiling Pt:334 °C (1013 hPa)
Melting Pt:155…157 °C
EINECS:215-926-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:155.0 to 158.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together