
2,7-Dibromo-9-fluorenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14348-75-5
:
Formula:C₁₃H₆Br₂O
MDL Number:MFCD00010790
:338 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:202.0 to 206.0 deg-C
Specification Test Results
Frequently Bought Together