Order Entry
Italy
ContactUsLinkComponent
cis-1,2-Cyclohexanediol ≥98.0% (by GC)
cis-1,2-Cyclohexanediol ≥98.0% (by GC)
Codice catalogo TCIAC1802-1G
Supplier:  TCI
CAS Number:  
undefined
cis-1,2-Cyclohexanediol ≥98.0% (by GC)
Codice catalogo TCIAC1802-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    98.0 to 102.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :116.16 g/mol
Boiling Pt:116°C
Melting Pt:98…101 °C
Density:1,183 g/cm³ (20 °C)
CAS Number:1792-81-0
Formula:C₆H₁₂O₂
MDL Number:MFCD00064944