
Cyclohexanecarbonyl chloride ≥98.0% (by GC)
: TCI
:2719-27-9
:
:C₇H₁₁ClO
:MFCD00001456
:185°C
:29 °C
:1,08 g/cm³ (20 °C)
:66°C
:220-322-7
:8,II
:146.62 g/mol
:3265
:GHS05
:Ambient
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Frequently Bought Together