Chroman-4-one ≥98.0% (by GC)
: TCI
:491-37-2
:
:C₉H₈O₂
:MFCD00006840
:148.16 g/mol
:128°C
:39 °C
:1,1291 g/cm³ (99.8 °C)
:Refrigerator
:207-736-3
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together