

4,4'-Biphenol ≥99.0% (by GC)
: TCI
:92-88-6
:
:C₁₂H₁₀O₂
:MFCD00002348
:186.21 g/mol
:355 °C (1013 hPa)
:281°C
:1,25 g/cm³ (20 °C)
:202-200-5
:3077
:9,III
:GHS07
:GHS09
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together