3'-(Trifluoromethyl)propiophenone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1533-03-5
:
Formula:C₁₀H₉F₃O
MDL Number:MFCD00039227
:202.18 g/mol
Boiling Pt:102°C
Flash Pt:108°C
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together