Order Entry
Italy
ContactUsLinkComponent
2-Phenylethyl isothiocyanate ≥97.0% (by GC)
Corrosive
Irritant
Health
Environment
2-Phenylethyl isothiocyanate ≥97.0% (by GC)
Catalog # TCIAP0986-10ML
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
2-Phenylethyl isothiocyanate ≥97.0% (by GC)
Catalog # TCIAP0986-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :163.24 g/mol
Merck Index:14,07226
Boiling Pt:126°C
Density:1,09 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-855-5
UN:1760
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:2257-09-2
Formula:C₆H₅CH₂CH₂NCS
MDL Number:MFCD00004821