
α,α,α,6-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:115029-23-7
:
Formula:C₈H₄F₄O₂
MDL Number:MFCD00061292
:208.11 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:98.0 to 104.0 deg-C
Specification Test Results
Frequently Bought Together