
3-Nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:121-51-7
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007435
Melting Pt:64 °C
Storage Temperature:Ambient
EINECS:204-476-2
UN:3261
ADR:8,III
:221.62 g/mol
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:61.0 to 64.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together