
2-Nitrobenzenesulphonyl chloride ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1694-92-4
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007430
Melting Pt:69.5…69.7 °C
Storage Temperature:Ambient
EINECS:216-907-1
UN:3261
ADR:8,II
:221.62 g/mol
:GHS05
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:64.0 to 70.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together