
2-Iodobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:63059-29-0
:
Formula:C₆H₄ClIO₂S
MDL Number:MFCD00833336
:302.52 g/mol
Boiling Pt:135°C
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:50.0 to 54.0 deg-C
Specification Test Results
Frequently Bought Together