
2,4-Difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13918-92-8
:
Formula:C₆H₃ClF₂O₂S
MDL Number:MFCD00042481
:212.6 g/mol
Boiling Pt:84°C
Density:1,581 g/cm³ (20 °C)
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together