Order Entry
Italy
ContactUsLinkComponent
4-(Methylthio)benzoic acid ≥97.0% (by GC, titration analysis)
Irritant
4-(Methylthio)benzoic acid ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1675-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-(Methylthio)benzoic acid ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1675-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    193.0 to 197.0 deg-C

Specification Test Results

About this item

 :
 :168.22 g/mol
EINECS:236-171-5
 :GHS07
CAS Number:13205-48-6
Formula:C₈H₈O₂S
MDL Number:MFCD00002555