
4-Aminobenzhydrazide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA1211-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:226.0 to 230.0 deg-C
- Solubility in hot Acetic acid:very faint turbidity
Specification Test Results
About this item
:
Boiling Pt:378 °C (1013 hPa)
Melting Pt:280…283 °C
:151.17 g/mol
EINECS:226-324-4
:GHS07
CAS Number:5351-17-7
Formula:C₇H₉N₃O
MDL Number:MFCD00007606