
2,4-Dichlorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
: TCI
:16271-33-3
:
:C₆H₃Cl₃O₂S
:MFCD00052712
:245.51 g/mol
:56 °C
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
Frequently Bought Together