Order Entry
Italy
ContactUsLinkComponent
 
Ethyl-2-chlorobenzoate ≥98.0% (by GC)
  TCIAC1671-25G
 :  TCI
 :  
Ethyl-2-chlorobenzoate ≥98.0% (by GC)
  TCIAC1671-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :184.62 g/mol
 :243°C
 :230-842-6
 :7335-25-3
 :C₉H₉ClO₂
 :MFCD00016338