
cis-2-Butene-1,4-diol ≥94.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6117-80-2
:
Formula:C₄H₈O₂
MDL Number:MFCD00002924
:88.11 g/mol
Merck Index:12,02390
Boiling Pt:235°C
Melting Pt:7°C
Density:1,07 g/cm³ (20 °C)
Flash Pt:128°C
EINECS:228-085-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 94.0 %
Specification Test Results
Frequently Bought Together