
3,7-Dimethyl-1-octanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-21-8
:
Formula:C₁₀H₂₂O
MDL Number:MFCD00002936
:158.28 g/mol
Boiling Pt:118°C
Density:0,849 g/cm³ (0 °C)
Flash Pt:110°C
EINECS:203-374-5
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together