

Pivalaldehyde (trimethylacetaldehyde) ≥95.0% (by GC)
Catalog # TCIAP0847-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:86.13 g/mol
Boiling Pt:75°C
Melting Pt:6 °C
Density:0,771 g/cm³ (20 °C)
Flash Pt:20°C
Storage Temperature:Refrigerator
EINECS:211-134-6
UN:1989
ADR:3,II
:GHS02
:GHS07
CAS Number:630-19-3
Formula:C₅H₁₀O
MDL Number:MFCD00006962