Order Entry
Italy
ContactUsLinkComponent
 
 
2,3-Dimercapto-1-propanol ≥95.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
2,3-Dimercapto-1-propanol ≥95.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Iodometric Titration):
    min. 95.0 %

 

Frequently Bought Together