


N,N-Dimethylformamide dimethyl acetal ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4637-24-5
:
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482
:119.16 g/mol
Boiling Pt:104°C
Density:0,893 g/cm³ (20 °C)
Flash Pt:5°C
EINECS:225-063-3
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together