

Methyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:333-27-7
:
Formula:C₂H₃F₃O₃S
MDL Number:MFCD00000409
:164.11 g/mol
Boiling Pt:99°C
Density:1,45 g/cm³ (20 °C)
Flash Pt:38°C
EINECS:206-371-7
UN:2920
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together