
Triisopropylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:80522-42-5
:
Formula:C₁₀H₂₁F₃O₃SSi
MDL Number:MFCD00009913
:306.42 g/mol
Boiling Pt:87°C
Density:1,136 g/cm³ (20 °C)
Flash Pt:100°C
UN:1760
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together