

4'-Aminoacetanilide ≥98.0% (by HPLC, titration analysis)
: TCI
:122-80-5
:
:C₈H₁₀N₂O
:MFCD00007853
:14,00411
:267 °C (1013 hPa)
:165 °C
:1,27 g/cm³ (20 °C)
:150.18 g/mol
:Ambient
:204-576-6
:GHS07
:GHS08
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:164.0 to 168.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together