Oxalic acid dihydrazide ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:996-98-5
:
Formula:C₂H₆N₄O₂
MDL Number:MFCD00007608
:118.1 g/mol
Melting Pt:240 °C
Density:1,458 g/cm³ (23 °C)
EINECS:213-640-2
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Potassium bromate Method):min. 96.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together