
2,2-Dichloroacetamide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD0306-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:98.0 to 101.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:127.96 g/mol
Boiling Pt:234°C
Melting Pt:98 °C
EINECS:211-674-2
:GHS07
Storage Temperature:Ambient
CAS Number:683-72-7
Formula:C₂H₃Cl₂NO
MDL Number:MFCD00008015