

Bromoacetyl bromide ≥98.0% (by GC, titration analysis)
: TCI
:598-21-0
:
:BrCH₂COOBr
:MFCD00000115
:201.85 g/mol
:150°C
:2,32 g/cm³ (20 °C)
:Ambient
:209-923-5
:2513
:8,II
:GHS05
:GHS08
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Frequently Bought Together