(S,S)-(+)-2,4-Pentanediol ≥98.0% (by GC)
TCIAP1412-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Optical purity (GC):min. 98.0 ee%
- Purity (GC):min. 98.0 %
- Melting point:45.0 to 50.0 deg-C
- Solubility in EtOH:almost transparency
:
:104.15 g/mol
:111 - 113 °C (19 -rr)
:45…48 °C
:Ambient
:72345-23-4
:C₅H₁₂O₂
:MFCD00064276