Order Entry
Italy
ContactUsLinkComponent
2,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
Irritant
2,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2343-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,4-Difluorocinnamic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2343-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    216.0 to 219.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :184.14 g/mol
 :GHS07
CAS Number:94977-52-3
Formula:C₉H₆F₂O₂
MDL Number:MFCD00010317