Diethyl fumarate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:623-91-6
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00064455
:172.18 g/mol
Boiling Pt:219°C
Melting Pt:2°C
Density:1,05 g/cm³ (20 °C)
Flash Pt:111°C
Storage Temperature:Ambient
EINECS:210-819-7
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together