
Streptomycin sulphate ≥95.0% (by total nitrogen and titration analysis)
: TCI
:3810-74-0
:
:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
:MFCD00037023
:1457.4 g/mol
:14,08826
:250 °C
:Refrigerator
:223-286-0
:6.1,III
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 95.0 % (calcd.on dried substance)
- Purity (Neutralization titration):min. 95.0 % (calcd.on dried substance)
- Specific rotation [a]20/D:-79.0 to -88.0 deg (C=1, H2O) (calcd.on dried substance)
- Drying loss:max. 5.0 %
Frequently Bought Together