Tween® 85 (Polysorbate) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:9005-70-3
:
MDL Number:MFCD01779641
Density:1,02 g/cm³ (20 °C)
Flash Pt:315°C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to Brown clear liquid to slightly cloudy liquid
- Water:max. 5.0 %
- Saponification Value:83.0 to 105.0
- Suitability for protein-analysis:to pass test
Specification Test Results
Frequently Bought Together