Tween® 40 (Polysorbate) for biochemical research
: TCI
:9005-66-7
:
:C₆₂H₁₂₂O₂₆
:MFCD00165345
:? 100 °C (1013 hPa)
:∼0.1 °C
:1,05 g/cm³ (20 °C)
:268°C
:Ambient
- Appearance:Light yellow to Yellow to Orange clear liquid to cloudy liquid
- Water:max. 5.0 %
- Saponification Value:41.0 to 55.0
- Acid Value:max. 4.0
- Suitability for protein-analysis:to pass test
Frequently Bought Together