Tween® 40 (Polysorbate)
Supplier: TCI
About this item
Synonyms: Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Boiling Pt:? 100 °C (1013 hPa)
- Melting Pt:∼0.1 °C
- Density:1,05 g/cm³ (20 °C)
- Flash Pt:≥ 149 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Boiling Pt:? 100 °C (1013 hPa)
- Melting Pt:∼0.1 °C
- Density:1,05 g/cm³ (20 °C)
- Flash Pt:≥ 149 °C
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid to cloudy liquid
- Water:max. 5.0 %
- Saponification Value:41.0 to 55.0
- Acid Value:max. 4.0



