
Tween® 80 (Polysorbate) for biochemical research
: TCI
:9005-65-6
:
:MFCD00082107
:14,07582
:? 100 °C (1013 hPa)
:–21 °C
:1,062 g/cm³ (20 °C)
:295°C
:Ambient
:500-019-9
:GHS07
- Appearance:Light orange to Yellow to Green clear liquid
- Water:max. 4.0 %
- Acid Value:max. 2.0
- Saponification Value:45.0 to 55.0
- Suitability for protein-analysis:to pass test
Frequently Bought Together