Isobutyl formate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Formic acid Isobutyl ester
CAS Number:542-55-2
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00003293
:102.13 g/mol
Merck Index:14,05143
Boiling Pt:98°C
Melting Pt:–96 °C
Density:0,90495 g/cm³ (0 °C)
Flash Pt:10°C
EINECS:208-818-1
UN:2393
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together