Order Entry
Italy
ContactUsLinkComponent
Diethyl disulphide ≥99.0% (by GC)
 
 
 
Diethyl disulphide ≥99.0% (by GC)
  TCIAD0486-25G
 :  TCI
 :  
undefined
 
 
 
Diethyl disulphide ≥99.0% (by GC)
  TCIAD0486-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :122.26 g/mol
 :154°C
 :–101.5 °C
 :0,984 g/cm³ (20 °C)
 :40°C
 :203-805-7
 :1993
 :3,III
 :GHS02
 :GHS07
 :GHS09
 :110-81-6
 :C₄H₁₀S₂
 :MFCD00009266