Metile oleato >60.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-62-9
:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578
:296.49 g/mol
Merck Index:14,06828
Boiling Pt:161°C
Melting Pt:–20°C
Density:0,88 g/cm³ (20 °C)
Flash Pt:163°C
Storage Temperature:Ambiente
EINECS:203-992-5
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 60.0 %
Specification Test Results
Related Information
Frequently Bought Together