DL-Arabinose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:147-81-9
:20235-19-2
Formula:C₅H₁₀O₅
MDL Number:MFCD00135867
:150.13 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:159…160 °C
Density:1,609 g/cm³ (17 °C)
Storage Temperature:Ambient
EINECS:205-699-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:157.0 to 162.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together