
5-Methylcaproic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:628-46-6
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00036494
:130.19 g/mol
Boiling Pt:216°C
Flash Pt:102°C
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together