
5-Aminopentan-1-ol ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2508-29-4
:
Formula:C₅H₁₃NO
MDL Number:MFCD00008237
Boiling Pt:222°C
Melting Pt:36 °C
Density:0,94 g/cm³ (20 °C)
Flash Pt:65°C
EINECS:219-718-2
ADR:8,III
:103.16 g/mol
UN:1759
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together