4-Methyl-1-pentanol ≥99.0% (by GC)
: TCI
: 4-Methylpentan-1-ol
:626-89-1
:
:C₆H₁₄O
:MFCD00002962
:102.18 g/mol
:152°C
:–70 °C
:0,84 g/cm³ (20 °C)
:57°C
:Ambient
:210-969-3
:1987
:3,III
:GHS02
:GHS07
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together