Order Entry
Italy
ContactUsLinkComponent
3-Methyl-4-heptanone ≥95.0% (by GC)
Flammable
3-Methyl-4-heptanone ≥95.0% (by GC)
Catalog # TCIAM1267-10ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
3-Methyl-4-heptanone ≥95.0% (by GC)
Catalog # TCIAM1267-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :128.21 g/mol
Boiling Pt:156°C
Flash Pt:38°C
EINECS:239-820-0
UN:1224
ADR:3,III
 :GHS02
CAS Number:15726-15-5
Formula:C₈H₁₆O
MDL Number:MFCD00048721