Order Entry
Italy
ContactUsLinkComponent
3-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Corrosive
Irritant
3-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Catalog # TCIAC0139-25G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
3-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Catalog # TCIAC0139-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:222°C
Melting Pt:44 °C
Density:1,367 g/cm³ (20 °C)
Flash Pt:117°C
Storage Temperature:Ambient
EINECS:210-552-6
UN:3265
ADR:8,II
 :175.01 g/mol
 :GHS05
 :GHS07
CAS Number:618-46-2
Formula:C₇H₄Cl₂O
MDL Number:MFCD00000671