
5-Bromo-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:146328-85-0
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD00143423
:219.01 g/mol
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:147.0 to 153.0 deg-C
Specification Test Results
Frequently Bought Together