
3-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
: TCI
:13472-59-8
:
:C₆H₆BrNO
:MFCD03095349
:93°C
:188.02 g/mol
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together