
3-(Trifluoromethoxy)benzaldehyde ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:52771-21-8
:
Formula:C₈H₅F₃O₂
MDL Number:MFCD00042406
:190.12 g/mol
Boiling Pt:83 - 86 °C (24 -rr)
Melting Pt:450 °C
Density:1,33 g/cm³ (20 °C)
Flash Pt:73°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together