Order Entry
Italy
ContactUsLinkComponent
3,9-Dodecadiyne ≥97.0% (by GC)
 
3,9-Dodecadiyne ≥97.0% (by GC)
  TCIAD1985-5ML
 :  TCI
 :  
undefined
 
3,9-Dodecadiyne ≥97.0% (by GC)
  TCIAD1985-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :162.27 g/mol
 :95°C
 :GHS07
 :61827-89-2
 :C₁₂H₁₈
 :MFCD00041637