Order Entry
Italy
ContactUsLinkComponent
3,3-Dimethylhexane ≥99.0% (by GC)
Flammable
Irritant
Health
Environment
3,3-Dimethylhexane ≥99.0% (by GC)
Catalog # TCIAD1227-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
3,3-Dimethylhexane ≥99.0% (by GC)
Catalog # TCIAD1227-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :114.23 g/mol
Boiling Pt:112°C
Melting Pt:-126°C
Melting Pt:–126°C
Density:0,7099 g/cm³ (20 °C)
Flash Pt:7°C
Storage Temperature:Ambient
EINECS:209-243-9
UN:1262
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:563-16-6
Formula:C₈H₁₈
MDL Number:MFCD00048729